missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone, 98+%

CAS: 7787-20-4 | C10H16O | 152.237 g/mol

$74.51 - $159.14

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1515118
View Documents
Thermo Scientific Chemicals
L1515118
50 g N/A
Only null left
 
AAL1515130
View Documents
Thermo Scientific Chemicals
L1515130
250 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-20-4
152.237
LHXDLQBQYFFVNW-GMSGAONNSA-N
3034206
CC1(C2CCC(C2)(C1=O)C)C
C10H16O
MFCD00151104
l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one

Specifications

7787-20-4
0.945
73°C (163°F)
1.463
50 g
l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
−52° (Neat)
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
3034206
≥98%
3°C to 5°C
192°C to 194°C
C10H16O
MFCD00151104
2042710
LHXDLQBQYFFVNW-GMSGAONNSA-N
CC1(C2CCC(C2)(C1=O)C)C
152.237
152.24
(-)-Fenchone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 232-107-5

RTECSNumber : RB7875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only