missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone, 98+%

Catalog No. AAL1515118
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1515118 50 g
AAL1515130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1515118 Supplier Thermo Scientific Chemicals Supplier No. L1515118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7787-20-4 | C10H16O | 152.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7787-20-4
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00151104
InChI Key LHXDLQBQYFFVNW-GMSGAONNSA-N
Synonym l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
PubChem CID 3034206
IUPAC Name (1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
SMILES CC1(C2CCC(C2)(C1=O)C)C

Specifications

Melting Point 3°C to 5°C
Density 0.945
Boiling Point 192°C to 194°C
Flash Point 73°C (163°F)
Refractive Index 1.463
Quantity 50 g
Beilstein 2042710
Optical Rotation −52° (Neat)
Formula Weight 152.24
Percent Purity ≥98%
Chemical Name or Material (-)-Fenchone
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 232-107-5
RTECSNumber RB7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only