Learn More
Eppendorf™ FemtoJet™ 4i Microinjector
The electronic microinjector, FemtoJet 4i, with its simple operation and wide range of functionality is ideally suited for injecting small to intermediate volumes of up to 100 pL into adherent cells and suspension cells for research purposes.
Supplier:  Eppendorf™ EPE5252000021
Encompass
Catalog No. E5252000021
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
The FemtoJet 4i with its built-in compressor forms a perfect team with Eppendorf micromanipulators, but it integrates just as easily with non-Eppendorf micromanipulator systems. The electronic connection between FemtoJet 4i and TransferMan™ 4r and InjectMan™ 4 guarantees simple, rapid and reproducible microinjections into cells.
  • The electronic microinjector, FemtoJet 4i, with its simple operation and wide range of functionality is ideally suited for injecting small to intermediate volumes of up to 100 pL into adherent cells and suspension cells for research purposes. .
  • Very simple and clearly arranged operation for routine tasks
  • Injection volume ranges from femtoliters to microliters
  • Inject and Clean function can be triggered right on the device using the included mouse or optional foot control
  • Programmable injection guarantees high reproducibility
  • Connection to TransferMan™ 4r, InjectMan™ 4 or InjectMan™ NI 2 enables semi-automatic microinjection
  • Small footprint
  • Can be combined with all conventional micromanipulation systems
  • Independent pressure supply via built-in compressor (only FemtoJet 4i)
  • External pressure supply guarantees the continuous availability of higher injection pressures - even with large volumes and very short intervals (only FemtoJet 4x)
Specifications
Specifications
50/60 Hz
Microinjection into small organisms and embryos in the early stages of development
Can be combined with all conventional micromanipulation systems
100/240 V
Product Suggestions
Product Suggestions