Learn More
Sartorius Female Luer Fittings
Catalog No. 14557111
Change view
Click to view available options
Material:
Tefzel
Catalog No. Material
14557111 Tefzel
1 options
Catalog No. 14557111 Supplier Sartorius Supplier No. 17002140
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Adapters to convert the Luer inlet and outlet connectors on any Sartobind™ Lab unit to the M6 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with M6 male connectors.
  • Adapters to convert the Luer inlet and outlet connectors on any Sartobind™ Lab unit to the M6 standard. This enables the operation of Sartobind™ Lab units on FPLC systems fitted with M6 male connectors.
  • 2 x M6 female to female Luer and 2 x M6 female to male Luer adapters
  • Black Tefzel
Specifications
Type Adapter
Color Black
Material Tefzel
Pressure 100 psi