missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Female Luer with 1/4 in. ID Barb - Natural Kynar - QC

Catalog No. 01000162
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01000162 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000162

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Precision engineered for smooth positive sealing. This standard luer is ISO 80369-7 compliant.

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Kynar PVDF
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Luer
Quantity 100 Pk.