missing translation for 'onlineSavingsMsg'
Learn More

Fehlings Solution 'A', Copper No. 1, Certified, 6.93% (w/v) ±0.30%, LabChem™

Catalog No. LC142001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer™
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC142001 500 mL Poly Bottle
LC142002 1 L Poly Bottle
LC142004 4 L Poly Bottle
LC142006 10 L Cubitainer™
LC142005 20 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC142001 Supplier LabChem Supplier No. LC142001
Only null left

Specifications

Chemical Name or Material Fehlings Solution 'A', Copper No. 1
Name Note Copper No. 1
CAS 7758-99-8, 7732-18-5
CAS Max % 93.07, 6.93
Linear Formula CuSO4·5H2O
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 249.68
Percent Purity 6.93% ±0.3% (w/v)
Grade Certified
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Fume
Color Blue
Quantity 500 mL
Physical Form Liquid
Show More Show Less