missing translation for 'onlineSavingsMsg'
Learn More

Fehling's Copper Solution (Fehling Solution A), Ricca Chemical

Catalog No. 301025
Change view
Click to view available options
Package: Cubitainer; Size: 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
301025 Package: Cubitainer; Size: 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 301025 Supplier Ricca Chemical Company Supplier No. RI30102.5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Determination of Reducing Sugars

Specifications

Chemical Name or Material Fehling's Copper Solution
Name Note Fehling Solution A
CAS 7758-99-8, 7732-18-5
Packaging Cubitainer
Quantity 10 L