Learn More
Instech Laboratories, Inc. Plastic feeding tubes, 13ga (purple) x 150mm, sterile
Catalog No. 5081038
Encompass
Plastic feeding tubes, 13ga x 150mm. 50 sterile pouches of 5. Polypropylene female luer (purple) and flexible polypropylene tube with soft elastomer tip. For rodent oral gavage. Disposable. Designed to minimize trauma to esophagus.
Catalog No. 5081038 Supplier Instech Laboratories, Inc. Supplier No. FTP13150
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plastic feeding tubes, 13ga x 150mm. 50 sterile pouches of 5. Polypropylene female luer (purple) and flexible polypropylene tube with soft elastomer tip. For rodent oral gavage. Disposable. Designed to minimize trauma to esophagus.