missing translation for 'onlineSavingsMsg'
Learn More

FD and C Yellow No. 6, Spectrum™ Chemical

Catalog No. p-7157469
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602641 100 g Poly Pail
18602642 500 g Poly Pail
18602643 2.5 kg Poly Pail
18602644 20 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602641 Supplier Spectrum Chemical Mfg Cor Supplier No. FD155100GM
Only null left

FD155, 2783-94-0, C16H10N2O7S2Na2

Spectrum™ Chemical FD and C Yellow No. 6

Chemical Identifiers

CAS 2783-94-0
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
InChI Key TXVRKNUZLYFDTJ-DDVLFWKVSA-L
IUPAC Name disodium (5E)-6-oxo-5-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-5,6-dihydronaphthalene-2-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(N\N=C2\C(=O)C=CC3=CC(=CC=C23)S([O-])(=O)=O)C=C1

Specifications

CAS Max % 100%
Percent Purity 87%
Grade Reagent
Packaging Poly Pail
Color Orange to Yellow
Quantity 100 g
Chemical Name or Material FD and C Yellow No. 6