missing translation for 'onlineSavingsMsg'
Learn More

FD and C Red No. 40, Spectrum™ Chemical

Catalog No. 18602635
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
20 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602635 25 g Poly Pail
18602631 100 g Poly Pail
18602632 500 g Poly Pail
18602633 5 kg Poly Pail
18602634 20 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602635 Supplier Spectrum Chemical Mfg Cor Supplier No. FD14025GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FD140, 25956-17-6, C18H14N2O8S2Na2

Spectrum™ Chemical FD and C Red No. 40 is used as a food dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 25956-17-6
Molecular Formula C18H14N2Na2O8S2
Molecular Weight (g/mol) 496.42
MDL Number MFCD00059526
InChI Key POXPUHKJGLHZEM-VIPPSAFOSA-L
IUPAC Name disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate
SMILES [Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Pail
Color Dark Red
Quantity 25 g
Percent Purity 85%
Grade Reagent
Chemical Name or Material FD and C Red No. 40