missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Red No. 40, Spectrum™ Chemical
FD140, 25956-17-6, C18H14N2O8S2Na2
Supplier: Spectrum Chemical Mfg Cor FD140100GM
| Quantity | 100 g |
|---|---|
| Packaging | Poly Pail |
Description
Spectrum™ Chemical FD and C Red No. 40 is used as a food dye and originally used as a replacement for amaranth as a food coloring. It looks like a dark red powder and often comes as a sodium salt. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 25956-17-6 | |
| 496.42 | |
| POXPUHKJGLHZEM-VIPPSAFOSA-L | |
| [Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O |
| C18H14N2Na2O8S2 | |
| MFCD00059526 | |
| disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate |
Specifications
| FD and C Red No. 40 | |
| 100% | |
| MFCD00059526 | |
| [Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O | |
| 496.42 | |
| Reagent | |
| Dark Red |
| 25956-17-6 | |
| C18H14N2Na2O8S2 | |
| POXPUHKJGLHZEM-VIPPSAFOSA-L | |
| disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate | |
| 85% | |
| Poly Pail | |
| 100 g |