missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ FBX09 Antibody Blocking Peptide

Catalog No. NB398424
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398424 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398424 Supplier R&D Systems™ Supplier No. NBP227364PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Blocking Peptide

Specifications

Gene ID (Entrez) 26268
Product Line Novus
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP2-27364. For further blocking peptide related protocol, click here .
Gene Alias Cross-immune reaction antigen 1, dJ341E18.2, F-box only protein 9, F-box protein 9, F-box protein Fbx9, FBX9DKFZp434°C0118, KIAA0936, NY-REN-57, Renal carcinoma antigen NY-REN-57, VCIA1
Gene Symbol FBXO9
Quantity 0.1 mg
Research Category Stem Cell Markers
Storage Buffer Peptide dissolved in dH20. Contains no BSA.