missing translation for 'onlineSavingsMsg'
Learn More

Fasudil monohydrochloride, 99+%

Catalog No. AAJ60751ME
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60751ME 500 mg
AAJ60751MD 250 mg
AAJ6075103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60751ME Supplier Thermo Scientific Chemicals Supplier No. J60751ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105628-07-7 | C14H18ClN3O2S | 327.827 g/mol

Fasudil monohydrochloride is used in the prevention and treating cardiovascular and cerebrovascular diseases. It is also used in the inhibition of proliferation of vascular smooth muscle cells. It serves as a selective Rho-associated protein kinase (ROCK) inhibitor. Further, it protects against methyl mercury-induced axonal degeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.

Notes
Incompatible with acids, bases and oxidizing agents.

Chemical Identifiers

CAS 105628-07-7
Molecular Formula C14H18ClN3O2S
Molecular Weight (g/mol) 327.827
MDL Number MFCD00943198
InChI Key LFVPBERIVUNMGV-UHFFFAOYSA-N
Synonym fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
PubChem CID 163751
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl

Specifications

Color White
Quantity 500 mg
Merck Index 14,3942
Solubility Information Soluble in water and dimethyl sulfoxide. Insoluble in ethanol.
Formula Weight 327.83
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Fasudil monohydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber HM4031166
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only