missing translation for 'onlineSavingsMsg'
Learn More

Fasudil Hydrochloride 98.0+%, TCI America™

Catalog No. F0839100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0839100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0839100MG Supplier TCI America Supplier No. F0839100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105628-07-7
Molecular Formula C14H18ClN3O2S
Molecular Weight (g/mol) 327.827
MDL Number MFCD00943198
InChI Key LFVPBERIVUNMGV-UHFFFAOYSA-N
Synonym fasudil hydrochloride, fasudil hcl, eril, at 877 hydrochloride, fasudil monohydrochloride, ha 1077 hydrochloride, unii-sq04n8s7br, 5-1,4-diazepan-1-yl sulfonyl isoquinoline hydrochloride, sq04n8s7br
PubChem CID 163751
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;hydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl

Specifications

Melting Point 222°C
Color White
Quantity 100 mg
Formula Weight 327.83
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fasudil Hydrochloride
RTECSNumber HM4033500
TSCA No