missing translation for 'onlineSavingsMsg'
Learn More

Fasudil, 98+%

Catalog No. AAJ63525MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63525MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63525MA Supplier Thermo Scientific Chemicals Supplier No. J63525MA
Only null left

CAS: 103745-39-7 | C14H17N3O2S | 291.369 g/mol

HA-1077 dihydrochloride has been shown to be a vasodilating agent as well as a potent inhibitor of Rock-2 (Rho-associated Kinase II), cGKI (cGMP-dependent protein kinase), PKA (cAMP-dependent protein kinase), MYLK, and CaMKII. Rho kinase signaling pathways are thought to cause invasion, migration, and proliferation of certain cells. Studies have shown this compound can also inhibit myosin light chain kinase in vitro and suppress cell rounding by myosin light chain phosphorylation. Mechanistic experiments report that HA-1077 inhibits vascular smooth muscle contraction by acting as a calcium channel protein inhibitor. HA-1077 dihydrochloride is an inhibitor of MSK1, PKC, PRK2 and Rsk-2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HA-1077 dihydrochloride has been shown to be a vasodilating agent as well as a potent inhibitor of Rock-2 (Rho-associated Kinase II), cGKI (cGMP-dependent protein kinase), PKA (cAMP-dependent protein kinase), MYLK, and CaMKII. Rho kinase signaling pathways are thought to cause invasion, migration, and proliferation of certain cells. Studies have shown this compound can also inhibit myosin light chain kinase in vitro and suppress cell rounding by myosin light chain phosphorylation. Mechanistic experiments report that HA-1077 inhibits vascular smooth muscle contraction by acting as a calcium channel protein inhibitor. HA-1077 dihydrochloride is an inhibitor of MSK1, PKC, PRK2 and Rsk-2.

Solubility
Soluble in water or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Specifications

CAS 103745-39-7
Color White
Molecular Formula C14H17N3O2S
Quantity 10 mg
MDL Number MFCD00866182
Merck Index 14,3942
Synonym fasudil, fasudil inn, 5-1,4-diazepane-1-sulfonyl isoquinoline, fasudilum inn-latin, 1-5-isoquinolinesulfonyl homopiperazine, unii-q0ch43pgxs, 1h-1,4-diazepine, hexahydro-1-5-isoquinolinylsulfonyl, 5-1,4-diazepan-1-ylsulfonyl isoquinoline, 5-isoquinolinesulfonyl homopiperazine, q0ch43pgxs
Solubility Information Soluble in water or DMSO
InChI Key NGOGFTYYXHNFQH-UHFFFAOYSA-N
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline
Molecular Weight (g/mol) 291.369
PubChem CID 3547
Formula Weight 327.83
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Fasudil
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only