missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOJO™ FAST TOWELS™ Hand and Surface Towels
Description
- Large, single textured wet shop towel
- All-in-one cleaning formula effective for cleaning hands, arms, tools and surfaces
- Contains skin moisturizers to help prevent dryness and irritation
- VOC-compliant formula
- For use anywhere heavy duty cleaning is required; no water needed
- Fresh citrus scent
- Effective on dirt, oil and grease
Specifications
Specifications
| Product Type | Hand Towels |
| Color | White |
| Form | Wipe |
| For Use With (Application) | Hand Towels |
| Packaging | Bucket |
| Quantity | 225 Count/Bucket, 2 Buckets/Pk. |