Learn More
Biotium Fast Probe Master Mix (5000 rxn)
Catalog No. NC1314152
Encompass
Fast Probe Master Mix is formulated for conducting real-time PCR using oligo-based probes such as TaqMan or MGB probes. The master mix contains buffer, dNTPs, MgCl2 and our superior hotstart enzyme Cheetah™ Taq. Cheetah™ Taq is a chemically modified hotstart Taq which can be fully activated in 2 minutes, thus permitting fast PCR. We offer two formats of probe master mix, without ROX and with ROX.For sensitive, affordable qPCR reactions we now recommend our new and improved Forget-Me-Not™ Universal Probe PCR Master Mix.TaqMan is aregistered trademark of Roche Diagnostics.
Catalog No. NC1314152 Supplier Biotium Supplier No. 310052
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Fast Probe Master Mix is formulated for conducting real-time PCR using oligo-based probes such as TaqMan or MGB probes. The master mix contains buffer, dNTPs, MgCl2 and our superior hotstart enzyme Cheetah™ Taq. Cheetah™ Taq is a chemically modified hotstart Taq which can be fully activated in 2 minutes, thus permitting fast PCR. We offer two formats of probe master mix, without ROX and with ROX.For sensitive, affordable qPCR reactions we now recommend our new and improved Forget-Me-Not™ Universal Probe PCR Master Mix.TaqMan is aregistered trademark of Roche Diagnostics.