missing translation for 'onlineSavingsMsg'
Learn More

GVS FAST™ Microarray Slides

Numéro de catalogue. p-4911406
Change view
Click to view available options
Includes:
1 Pad
2 Pads
8 Pads
Dimensions (LxW):
6 x 6 mm
20 x 20 mm
20 x 51 mm
20 x 60 mm
Quantity:
10/Pk.
20/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Includes Dimensions (LxW) Quantity
09977709 1 Pad 20 x 51 mm 20/Pk.
09977710 1 Pad 20 x 60 mm 20/Pk.
09977711 2 Pads 20 x 20 mm 10/Pk.
09977712 8 Pads 6 x 6 mm 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 09977709 Fournisseur GVS Code fournisseur 10484182
Il en reste null