missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fast Green FCF 85.0+%, TCI America™
$45.95 - $110.30
Chemical Identifiers
| CAS | 2353-45-9 |
|---|---|
| Molecular Formula | C37H34N2Na2O10S3 |
| Molecular Weight (g/mol) | 808.84 |
| MDL Number | MFCD00013053 |
| InChI Key | XJBPDZVCYTYRIN-UHFFFAOYSA-L |
| PubChem CID | 16887 |
| IUPAC Name | disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium |
| SMILES | [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O |
Chemical Identifiers
| 2353-45-9 | |
| 808.84 | |
| XJBPDZVCYTYRIN-UHFFFAOYSA-L | |
| disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium |
| C37H34N2Na2O10S3 | |
| MFCD00013053 | |
| 16887 | |
| [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O |
Specifications
| 2353-45-9 | |
| MFCD00013053 | |
| [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O | |
| 808.84 | |
| 808.84 | |
| Crystal/Powder |
| C37H34N2Na2O10S3 | |
| XJBPDZVCYTYRIN-UHFFFAOYSA-L | |
| disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium | |
| 16887 | |
| ≥85.0% (HPLC,E) | |
| Fast Green FCF |
Safety and Handling
EINECSNumber : (5)-5228
RTECSNumber : BQ4425000
TSCA : Yes