missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fast Green FCF 85.0+%, TCI America™
Specifications
Specifications
| Formula Weight | 808.84 |
| Percent Purity | ≥85.0% (HPLC,E) |
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Fast Green FCF [for Biochemical Research] |
Safety and Handling
| EINECSNumber | (5)-5228 |
| RTECSNumber | BQ4425000 |
| TSCA | Yes |