Learn More
Waters Limited Fast Fruit Juice Guard-Pak Insert, 10/pk
Catalog No. 50785074
Encompass
The Fast Fruit Juice Guard-Pak Insert is used to extend and prolong the lifetime for the Fast Fruit Juice Column by removing chemical and particulate contaminants from the mobile phase stream.
Catalog No. 50785074 Supplier Waters Limited Supplier No. WAT015207
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Fast Fruit Juice Guard-Pak Insert is used to extend and prolong the lifetime for the Fast Fruit Juice Column by removing chemical and particulate contaminants from the mobile phase stream.