missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB base

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol

$94.08 - $225.96

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6245614
View Documents
Thermo Scientific Chemicals
J6245614
25 g
Each for $94.08
Only null left
 
AAJ6245622
View Documents
Thermo Scientific Chemicals
J6245622
100 g
Each for $225.96
Only null left
 
Description

Description

C.I. 37175

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 37175
Specifications

Chemical Identifiers

120-00-3
300.358
CNXZLZNEIYFZGU-UHFFFAOYSA-N
67108
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
C17H20N2O3
MFCD00009091
C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide
N-(4-amino-2,5-diethoxyphenyl)benzamide

Specifications

95°C to 105°C
C17H20N2O3
3416889
CNXZLZNEIYFZGU-UHFFFAOYSA-N
N-(4-amino-2,5-diethoxyphenyl)benzamide
67108
Brown to Gray
Powder
120-00-3
MFCD00009091
C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
300.358
300.35
25 g
Fast Blue BB base
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H300-H315-H319-H335-H500

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 204-363-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only