missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB base

Catalog No. AAJ6245614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6245614 25 g
AAJ6245622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6245614 Supplier Thermo Scientific Chemicals Supplier No. J6245614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol

C.I. 37175

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C.I. 37175

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym C.I. 37175; 4'-Amino-2',5'-diethoxybenzanilide
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2

Specifications

Melting Point 95°C to 105°C
Beilstein 3416889
Formula Weight 300.35
Color Brown to Gray
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fast Blue BB base
Hazard Category H300-H315-H319-H335-H500
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 204-363-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only