missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB, 98%, pure

Catalog No. AC191570250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191570250 25 g Glass bottle
AC191571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC191570250 Supplier Thermo Scientific Chemicals Supplier No. 191570250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-00-3 | C17H20N2O3 | 300.358 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym 4'-Amino-2', 5'-diethoxybenzanilide, Azoic Diazo No.20, C.I. 37175
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2

Specifications

Melting Point 98.0°C to 100.0°C
Assay Percent Range 98%
Beilstein 13, III, 2170
Infrared Spectrum Authentic
Formula Weight 300.35
Percent Purity 98%
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Brown to Gray
Quantity 25 g
Physical Form Powder or Chunks
Chemical Name or Material Fast blue BB, 98%
Show More Show Less
Hazard Statement GHS P Statement
Fatal if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-363-8
TSCA TSCA

RUO – Research Use Only