missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fast Black K Salt, TCI America™
Supplier: TCI America F009225G
Specifications
| Fast Black K Salt | |
| 64071-86-9 | |
| MFCD00078471 | |
| XTWKPSDLMBAAGG-UHFFFAOYSA-K | |
| zinc;2,5-dimethoxy-4-[(4-nitrophenyl)diazenyl]benzenediazonium;trichloride | |
| 91972136 | |
| 25 g |
| 150°C | |
| C14H12Cl3N5O4Zn | |
| 2,5-Dimethoxy-4-(4-nitrophenylazo)benzenediazonium Chloride Zinc Chloride | |
| COC1=CC(=C(C=C1N=NC2=CC=C(C=C2)[N+](=O)[O-])OC)[N+]#N.[Cl-].[Cl-].[Cl-].[Zn+2] | |
| 486.011 | |
| 417.87 | |
| Crystal/Powder |
Chemical Identifiers
| 64071-86-9 | |
| 486.011 | |
| XTWKPSDLMBAAGG-UHFFFAOYSA-K | |
| 91972136 | |
| COC1=CC(=C(C=C1N=NC2=CC=C(C=C2)[N+](=O)[O-])OC)[N+]#N.[Cl-].[Cl-].[Cl-].[Zn+2] |
| C14H12Cl3N5O4Zn | |
| MFCD00078471 | |
| 2,5-Dimethoxy-4-(4-nitrophenylazo)benzenediazonium Chloride Zinc Chloride | |
| zinc;2,5-dimethoxy-4-[(4-nitrophenyl)diazenyl]benzenediazonium;trichloride |
Safety and Handling
EINECSNumber : (5)-2322
TSCA : Yes