Learn More
ACROBiosystems Inc Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein;HEK293;1MG;FAS,FAS,ALPS1A,APO-1,APO1,APT1,APT-1,CD95,CD-95,FAS1,FAS-1,FASTM,TNFRSF6,TNFRSF-6,FasR ,Fas-R
Catalog No. 501657579
Encompass
Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein;1MG;Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 173 (Accession # AAH12479.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.2 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501657579 Supplier ACROBiosystems Inc Supplier No. FASH52291MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human Fas / TNFRSF6 / CD95 Protein;1MG;Human Fas, His Tag (FAS-H5229) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Asn 173 (Accession # AAH12479.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.2 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.