missing translation for 'onlineSavingsMsg'
Learn More

Faropenem sodium hydrate, Thermo Scientific Chemicals

Catalog No. AC464962500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464962500 250 mg
AC464960010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464962500 Supplier Thermo Scientific Chemicals Supplier No. 464962500
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122547-49-3
Molecular Formula C12H14NNaO5S·xH2O
Molecular Weight (g/mol) 307.3

Specifications

Quantity 250 mg
Formula Weight 307.3
Chemical Name or Material Faropenem sodium hydrate

RUO – Research Use Only