missing translation for 'onlineSavingsMsg'
Learn More

Farnesylacetone, mixture of isomers, 97%

Catalog No. AAA1923430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1923430 250 g
AAA1923418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1923430 Supplier Thermo Scientific Chemicals Supplier No. A1923430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 762-29-8 | C18H30O | 262.437 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 762-29-8
Molecular Formula C18H30O
Molecular Weight (g/mol) 262.437
MDL Number MFCD00036517
InChI Key LTUMRKDLVGQMJU-IUBLYSDUSA-N
Synonym farnesyl acetone, 5e,9e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, farnesylacetone, e,e-farnesylacetone, unii-3s0g4n267h, e,e-6,10,14-trimethylpentadeca-5,9,13-trien-2-one, trans,trans-farnesylacetone, 5,9,13-pentadecatrien-2-one, 6,10,14-trimethyl-, e,e, 2,6,10-trimethyl-2,6,10-pentadecatrien-14-one, 5,9,13-pentadecatriene-2-one, 6,10,14-trimethyl
PubChem CID 1711945
ChEBI CHEBI:67252
IUPAC Name (5E,9E)-6,10,14-trimethylpentadeca-5,9,13-trien-2-one
SMILES CC(=CCCC(=CCCC(=CCCC(=O)C)C)C)C

Specifications

Density 0.88
Flash Point >110°C (230°F)
Odor Pleasant
Refractive Index 1.481
Quantity 250 g
Beilstein 1781239
Formula Weight 262.44
Percent Purity 97%
Chemical Name or Material Farnesylacetone, mixture of isomers
EINECSNumber 212-097-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only