missing translation for 'onlineSavingsMsg'
Learn More

Farnesyl acetate, mixture of isomers, 96%

Catalog No. AAA1977814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1977814 25 g
AAA1977822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1977814 Supplier Thermo Scientific Chemicals Supplier No. A1977814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 29548-30-9 | C17H28O2 | 264.409 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29548-30-9
Molecular Formula C17H28O2
Molecular Weight (g/mol) 264.409
MDL Number MFCD00036516
InChI Key ZGIGZINMAOQWLX-NCZFFCEISA-N
Synonym farnesyl acetate, farnesol acetate, all-trans-farnesyl acetate, unii-d5zj1foc2i, trans,trans-farnesyl acetate, d5zj1foc2i, 3,7,11-trimethyl-2,6,10-dodecatrienyl acetate, 2e,6e-farnesyl acetate, 2,6,10-dodecatrien-1-ol, 3,7,11-trimethyl-, acetate, e,e, 2e,6e-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
PubChem CID 638500
IUPAC Name [(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienyl] acetate
SMILES CC(=CCCC(=CCCC(=CCOC(=O)C)C)C)C

Specifications

Density 0.91
Flash Point 95°C (203°F)
Refractive Index 1.477
Quantity 25 g
Beilstein 1784823
Formula Weight 264.41
Percent Purity 96%
Chemical Name or Material Farnesyl acetate, mixture of isomers
EINECSNumber 249-689-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only