missing translation for 'onlineSavingsMsg'
Learn More

Fargesin 98.0+%, TCI America™

Catalog No. p-7130094
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F118810MG 10 mg
F118850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F118810MG Supplier TCI America Supplier No. F118810MG
Only null left

Chemical Identifiers

CAS 31008-19-2
Molecular Formula C21H22O6
Molecular Weight (g/mol) 370.40
MDL Number MFCD07781423
InChI Key AWOGQCSIVCQXBT-UHFFFAOYNA-N
Synonym rel-5-[(1R,3aS,4S,6aS)-4-(3,4-Dimethoxyphenyl)tetrahydro-1H,3H-furo[3,4-c]furan-1-yl]-1,3-benzodioxole
PubChem CID 92966415
IUPAC Name 5-[4-(3,4-dimethoxyphenyl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
SMILES COC1=C(OC)C=C(C=C1)C1OCC2C1COC2C1=CC2=C(OCO2)C=C1

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 10 mg
Formula Weight 370.40
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fargesin