missing translation for 'onlineSavingsMsg'
Learn More

Famciclovir 98.0+%, TCI America™

Catalog No. f0842500mg
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0842500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0842500MG Supplier TCI America Supplier No. F0842500MG
Only null left

Chemical Identifiers

CAS 104227-87-4
Molecular Formula C14H19N5O4
Molecular Weight (g/mol) 321.337
MDL Number MFCD00866964
InChI Key GGXKWVWZWMLJEH-UHFFFAOYSA-N
Synonym 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate
PubChem CID 3324
ChEBI CHEBI:4974
IUPAC Name [2-(acetyloxymethyl)-4-(2-aminopurin-9-yl)butyl] acetate
SMILES CC(=O)OCC(CCN1C=NC2=CN=C(N=C21)N)COC(=O)C

Specifications

Melting Point 105°C
Color White
Quantity 500 mg
Formula Weight 321.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Famciclovir
RTECSNumber TY3164000
TSCA No
Recommended Storage Refrigerator