Learn More
Excelta™ Excelta™ High Precision (Three Star) Single Edge Carbon Steel Razor Blades
Catalog No. 18100970
Change view
Click to view available options
Quantity:
10/Pk.
115/Pk.
Blade Material:
2 Facet Carbon Steel
3 Facet Carbon Steel
Carbon Steel
Packaging Type:
Individually Packaged
Unwrapped
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Blade Material Packaging Type
18100970 10/Pk. 2 Facet Carbon Steel Individually Packaged
18100971 115/Pk. - Unwrapped
18100972 115/Pk. Carbon Steel -
18100973 10/Pk. 3 Facet Carbon Steel Individually Packaged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18100970 Supplier Excelta™ Supplier No. 1761
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Three star carbon steel blades are offered individually wrapped, uwrapped and unwrapped and washed
  • Crafted of carbon steel
  • .009 single edge
  • 2 or 3 facet
  • Individually wrapped, unwrapped or unwrapped and washed
Specifications
Blade Material 2 Facet Carbon Steel
Quantity 10/Pk.
Packaging Type Individually Packaged