missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific False Bottom Tubes for Automation

For use as secondary tube for barcoding on automated analyzers

 View more versions of this product

Catalog No. 361016882


Only null left

Description

Description

For use as secondary tube for barcoding on automated analyzers

 

  • Made of Polypropylene
  • Dimensions: 16 x 97mm
  • Capacity: 5mL
  • Screw Caps are sold separately (See 21-200-231, -232)

 

Specifications

Chemical Identifiers

Polypropylene
False Bottom Tube

Specifications

Natural
97 mm
Secondary tube for barcoding on automated analyzers
16 mm
False Bottom Tube
5 mL
Polypropylene
1500/Cs.
Not Included
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions