Learn More
Fisherbrand™ False Bottom Tubes
Catalog No. 22170182
Change view
Click to view available options
Volume (Metric):
5 mL
Catalog No. Volume (Metric)
22170182 5 mL
1 options
Catalog No. 22170182 Supplier Fisherbrand™ Supplier No. 5527PLFH
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
False bottom tubes are designed for use as a secondary tube for barcoding on automated analyzers.
  • Disposable
Specifications
Volume (Metric) 5 mL
Sterility Non-sterile
Closure Type Screw Cap
Color Natural
Material Polypropylene