missing translation for 'onlineSavingsMsg'
Learn More

Starplex™ Scientific False Bottom Analyzer Tube

Catalog No. p-192732
Change view
Click to view available options
Color:
Clear
White
Product Type:
False Bottom Analyzer Vial
Screw Cap
Material:
Plastic
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Material
14375439 Clear False Bottom Analyzer Vial Polypropylene
361020455 White Screw Cap Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14375439
Only null left

Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes

  • Ideal for primary or secondary sampling as well as storage
  • No need for micro sample cups and liquid transfer devices
  • Accept bar codes and fit into most high volume analyzer racks
  • Compatible with Hitachi™, Olympus™, Corning™, Ortho (Kodak™), VITROS™ (750 and 250 series), and Beckman™ clinical chemistry analyzer

Order Info

Plug caps sold separately.

Specifications

Material Polypropylene
Color Clear
Product Type False Bottom Analyzer Vial
Sterility Non-sterile
Volume (Metric) 5 mL
For Use With (Application) Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes