missing translation for 'onlineSavingsMsg'
Learn More

Starplex™ Scientific False Bottom Analyzer Tube

Catalog No. 361020455
Change view
Click to view available options
Color:
Clear
White
Product Type:
False Bottom Analyzer Vial
Screw Cap
Material:
Plastic
Polypropylene
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Material
361020455 White Screw Cap Plastic
14375439 Clear False Bottom Analyzer Vial Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361020455 Supplier Starplex™ Scientific Supplier No. V9201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for high-volume analyzer aspiration probes incapable of reaching the bottoms of larger sample tubes

  • Ideal for primary or secondary sampling as well as storage
  • No need for micro sample cups and liquid transfer devices
  • Accept bar codes and fit into most high volume analyzer racks
  • Compatible with Hitachi™, Olympus™, Corning™, Ortho (Kodak™), VITROS™ (750 and 250 series), and Beckman™ clinical chemistry analyzer

Order Info

Plug caps sold separately.

Specifications

Color White
Product Type Screw Cap
Sterility Non-sterile
Material Plastic
Closure Type Screw
For Use With (Equipment) Starplex false bottom auto analyzer tube