Learn More
Falcon™ Cell Scrapers
Designed to provide maximum accessibility to growth surfaces. Falcon™ Cell Scrapers provide superior scraping ability with dishes, plates, and flasks.
Supplier: Falcon™ 353086
Description
- The improved Falcon Cell Scrapers have been thoughtfully designed to provide maximum accessibility to the growth surfaces in a variety of culture vessels
- Cross-ribbed polystyrene handle provides greater rigidity to ensure better control while scraping cells
- Flexibility of the joint between the blade and handle improves ease of access into the neck of a flask or roller bottle Highly compliant thermoplastic elastomer (TPE) blade pivots to provide multiple angles to remove cells from the entire growth surface
- Available in four sizes—choose the blade and handle length to best accommodate your needs
- Supplied individually in peel-open, medical-style packaging for sterile presentation
Order Info
Presterilized and individually wrapped in peel-open, medical-style packaging
Chemical Identifiers
| 1/Pk. | |
| Cell Scraper | |
| Sterile |
| 100/Cs. | |
| Compliant TPE blade and rigid, cross-ribbed polystyrene handle |
Specifications
| 0.7 in. | |
| For a variety of cell culture vessels, large flasks | |
| 25 cm | |
| 100/Cs. | |
| Compliant TPE blade and rigid, cross-ribbed polystyrene handle |
| 1.8 cm | |
| 10 in. | |
| 1/Pk. | |
| Cell Scraper | |
| Sterile |