missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Cell Culture Multi Flasks
GREENER_CHOICE

Catalog No. 1482696
Change view
Click to view available options
Culture Area:
525 cm2
875 cm2
Surface Coating:
3 Layer TC-Treated
TC-Treated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Surface Coating
1482696 875 cm2 TC-Treated
1482695 525 cm2 3 Layer TC-Treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1482696 Supplier Falcon™ Supplier No. 353144
Get Up to $2,500 in Products with Purchase
View all promotions
Edge

Grow cells faster and easier, increasing productivity in cell culture workflow. Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.

Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.

  • Even distribution of media across all layers for homogeneous cell growth
  • Ability to mix cells and reagents in the Falcon Multi-Flask saves time
  • Flexible design allows to pour or aspirate/recover cells using a pipet
  • Consistent surface treatment for predictable scale-up
  • Lot number printed on each flask for traceability
  • Manufactured in compliance to cGMP standards

Specifications

Diameter (Metric) Outer Neck 32 mm
Material Polystyrene
Height (English) 3.35 in.
Length (English) With cap: 7.8 in.
Neck Style Straight
No. per Case 8/Cs.
No. per Pack 1/Pk.
Product Type Cell Culture Multi-Flask
Shape Rectangular
Surface Coating TC-Treated
Width (English) 5 in.
Width (Metric) 127 mm
Disposable Single-use
Graduated Yes
Sterility Sterile
Bottom Shape Rectangular
Closure Type Vented
Culture Area 875 cm2
Surface Treatment Tissue Culture-treated
Volume (Metric) Working >5 to 50 mL/Layer
Tray Layers 5
Show More Show Less

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.