missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ FluoroBlok™96 Well HTS Insert Systems
GREENER_CHOICE

Catalog No. 08771008
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Well Shape
08771008 Flat Bottom (F-well)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08771008 Supplier Falcon™ Supplier No. 353928
Get Up to $2,500 in Products with Purchase
View all promotions

Simplify detection of migrating fluorescently labeled cells. Falcon™ FluoroBlok™96 Well HTS Insert Systems are designed with a unique light-tight PET membrane that blocks the transmission light within the range of 400 to 700nm.

96 square well, flat-bottom microplate
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • Specifications

    Format Standard
    Lid With Lid
    Culture Area 0.08 cm2
    Array 8 x 12
    For Use With (Equipment) FluoroBlok™ 96 Well Multiwell HTS Insert System
    Material Polystyrene
    No. of Wells 96
    Sterility Sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    Includes 96 square well, flat bottom plate
    No. per Case 5/Cs.
    No. per Pack 5/Pk.
    Quantity 5/Cs.
    Show More Show Less