missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Professional Faceshields
Description
- Protects against infrared (IR) and ultraviolet radiation (UVA / UVB) during tasks like light cutting and brazing
- Made of durable polycarbonate material
- Resusable (depending on the task)
- Designed for use in light industrial manufacturing, general splash protection, landscaping and food processing
- D3 rating for splash protection
- Compatible with the Jackson Safety Hard Hat Interchange System
- Snaps into place on compatible head gear and hard hat systems – no special tools required
- When used with any Jackson Safety Brand Headgear, this polycarbonate faceshield meets the performance standards of ANSI Z87.1+ for high-impact protection and is CSA Z94.3 compliant (when used with compatible Jackson Safety headgear)
Specifications
Specifications
| Width (English) | 15.5 in. |
| Lens Tint | Clear, IR Filter Shade 5.0 |
| Product Type | Faceshield |
| Length (English) | 8 in. |
| Lens Material | Polycarbonate |