Learn More
3M™ Faceshield Cover, H-111-100
Keeps the lens on the 3M™ Hoods H-Series clean. When vision becomes obstructed, the cover can be peeled off leaving a clean shield.
Supplier:  3M™ 7100138629
Catalog No. 29-663-297
Only null left
Explore available promotions
Description
Description
  • For use with 3M™ Hoods H-Series
  • Can be removed easily. Helps to keep your hood lens clean
  • Use compatible 3M™ Versaflo™ Headgear for your 3M™ Products
Specifications
Specifications
Clear
Faceshield Cover
1 Ea.
Product Suggestions
Product Suggestions