Learn More
United Scientific™ Face Shield
Description
Safe:
- Splash protection
- Covers entire face
- Lightweight
- Wear with our without goggles
- Non-sterile
- Anti-fog
- Produced in an FDA registered factory
- Comfortable:
- Soft elastic band
- Padded headband
- Clear PET shield
- Easy:
- Individually wrapped
- Disposable or can be sanitized
- Simple to put on and take off
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | None |
| Product Type | Faceshield |
| Material | PET |
| Disposable | Reusable |
| Lens Material | PET |
| Frame Material | Plastic |
| UV Protection | None |