missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ Respiratory Face Shield Assembly

Catalog No. 19121504
Change view
Click to view available options
Includes:
Comfort Faceseal
Flame Resistant Faceseal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19121504 Comfort Faceseal
19121506 Flame Resistant Faceseal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19121504 Supplier 3M™ Supplier No. 7100086942

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3M™ Versaflo™ respiratory face shield assembly is a lightweight, comfortable PAPR head top that combines a respirator and face shield, with the option of adding ear muffs for hearing protection.

  • Complete, fully assembled PAPR head top allows for use right out of the box
  • Flexible application, use with any 3M Versaflo system
  • Hearing protection compatible with M-985 ear muffs
  • May be a comfortable PAPR alternative to negative pressure respirators
  • May be a comfortable alternative to standard face shields
  • Easy to use design with ratchet suspension and liftable visor

Specifications

Color Gray
Product Type Headgear
For Use With (Application) Sanding, cleaning, demolition, grinding
For Use With (Equipment) 3M™ Versaflo™ PAPR and Supplied Air Systems
Includes Comfort Faceseal
Model M-206N
Suspension Type Ratchet