Learn More
Epredia™ Series 115 Laser Slotted Cassettes with Lids
Manufactured specifically for laser printing, the Epredia™ Series 115 Slotted Cassettes come with separate lids and are available in multiple colors.
$741.02 - $741.09
Specifications
Closure Included Yes
Quantity 40 Cassettes/Taped Stack, 25 Stacks/Cs.
Pore Style Slots
Closure Type Separate Lid with Tab
Product Type Cassette
View More Specs
Products 10
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
1150110LM
View Documents Promotion Details
Epredia™
1150110LM
White
Case of 1000 Each for $741.02
Only null left
1150210LM
View Documents Promotion Details
Epredia™
1150210LM
Yellow
Case of 1000 Each for $741.02
Only null left
1150310LM
View Documents Promotion Details
Epredia™
1150310LM
Blue
Case of 1000 Each for $741.02
Only null left
1150410LM
View Documents Promotion Details
Epredia™
1150410LM
Pink
Case of 1000 Each for $741.02
Only null left
1150510LM
View Documents Promotion Details
Epredia™
1150510LM
Green
Case of 1000 Each for $741.02
Only null left
1150610LM
View Documents Promotion Details
Epredia™
1150610LM
Orange
Case of 1000 Each for $741.02
Only null left
1150710LM
View Documents Promotion Details
Epredia™
1150710LM
Gray
Case of 1000 Each for $741.02
Only null left
1150810LM
View Documents Promotion Details
Epredia™
1150810LM
Aqua
Case of 1000 Each for $741.02
Only null left
1150910LM
View Documents Promotion Details
Epredia™
1150910LM
Lilac
Case of 1000 Each for $741.09
Only null left
1151210LM
View Documents Promotion Details
Epredia™
1151210LM
Red
Case of 1000 Each for $741.09
Only null left
Description
Description
  • Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
  • Contain a special constituent chemical compound that carbonizes when exposed to the laser beam
  • Each style comes in different color with color-matched lids
  • Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
  • Comes in taped stacks of 40 cassettes; 25 stacks per case for a total of 1000 cassettes with separate lids
  • The Epredia™ Laser Cassette is not compatible with thermal tape style printers
Specifications
Specifications
Yes
Slots
Cassette
40 Cassettes/Taped Stack, 25 Stacks/Cs.
Separate Lid with Tab