Learn More
Epredia™ Series 110 Laser Slotted with Front Tab Cassettes with Lids
Manufactured specifically for laser printing, the Epredia™ Series 110 Slotted Cassettes come with separate lids with a tab in the front for easy opening and closing.
Supplier: Epredia™ 1100910LM
Description
- Epredia™ Laser Cassettes designed for use on both the NOVA™ and VEGA™ Laser Printers
- Contain a special constituent chemical compound that carbonizes when exposed to the laser beam
- Each style comes in different colors with color-matched lids
- Each cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Epredia™ cassette hoppers for printing
- Comes in taped stacks of 40 cassettes; 25 stacks per case (for a total of 1000 cassettes with separate lids)
- The Epredia™ Laser Cassette is not compatible with thermal tape style printers
Specifications
| Slots | |
| 40 Cassettes/Taped Stack, 25 Stacks/Cs. | |
| Lilac |
| Yes | |
| Separate Lid with Tab | |
| Cassette |