Learn More
Beckman Coulter F2402H ROTOR, FIXED ANGLE, ALU
Catalog No. NC1495060
Encompass
F2402H Rotor, Fixed Angle, Aluminum, Biocontainment, 24 x 2.0 mL, 18,000 rpm, 29,756 x g
Catalog No. NC1495060 Supplier Beckman Coulter Supplier No. 361171
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
F2402H Rotor, Fixed Angle, Aluminum, Biocontainment, 24 x 2.0 mL, 18,000 rpm, 29,756 x g