Learn More
Innovatek EZTest - Ethylene Oxide Gas B. atrophaeus
Innovatek EZTest - Ethylene Oxide Gas B. atrophaeus is an easy-to-use test for monitoring sterilization
Supplier:  Innovatek 4140EZG/6
 View more versions of this product
Catalog No. 29801070
Only null left
Description
Description
  • EZTest is a self-contained biological indicator (BI) for monitoring sterilization
  • Ethylene Oxide Gas - B. atrophaeus Self-Contained Biological Indicator
  • Bacterial spores on a carrier material packaged within a small, thermoplastic culture tube
  • Sealed-glass ampoule of specially formulated soybean casein digest culture medium
  • Color indicator turns noticeably yellow when growth occurs
  • Easy-to-use in-office; no laboratory transfer needed
  • Dramatic visual results within 48 hours
Specifications
Chemical Identifiers
Yellow Indicator
EZTest™
48 hr. at 37 °C
Monitoring Sterilization
37°C
106
Specifications
Yellow Indicator
EZTest™
48 hr. at 37 °C
100 Tests/Pk.
Monitoring Sterilization
37°C
106
Product Suggestions
Product Suggestions