missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EZFluor™ 1-step Fluorescent Protein Gel Stain

Catalog No. SCT1451L
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCT1451L 1 L
SCT1454L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SCT1451L Supplier MilliporeSigma™ Supplier No. SCT1451L

Fluorescent protein gel stain that requires only a single 5 to 30 minute staining step without fixation.

  • Ready-to-use luminescent protein gel stain
  • Requires only a single 5 to 30 minute staining step without fixation
  • Destaining is optional
  • Offers safer handling and ease of disposal, because it is an aqueous-based solution that does not contain hazardous methanol or acetic acid
  • Passed environmental toxicity testing and is classified as non-hazardous to the environment under CCR Title 22 regulations
  • Can detect as little as approximately 1 to 10 ng of protein per band depending on the staining method used
  • Staining is fully compatible with mass spectrometry and Edman-based sequencing
  • Absorbance: 288 nm
  • Emission: 603 nm

Specifications

Description Absorbance: 288 nm; Emission: 603 nm
Detection Fluorescence
Form Liquid
Quantity 1 L
Storage Requirements Store at 2 to 8°C, protect from light