Learn More
Foxx Life Sciences EZBio Rigid Reservoir for Tangential Flow Filtration (TFF) System, PP, 2L HD Welded Bottle, 53B, VersaCap with Rigid tubing 3x Mini TC on 1/4""ID x 3/8""OD Tubing, Non-Sterile, 1/CS
GREENER_CHOICE
Catalog No. 5000332755
Encompass
The EZBio SUT Rigid Reservoir System is a single-use rigid bioprocess carboy and bottle solution for fluid handling, supporting upstream and downstream processes. It provides a sterile, closed, scalable platform with integrated rigid tubing for secure, kink-free transfer of buffers, media, process fluids.
Available in 1L115L volumes (PP bottles/carboys), it features VersaCap technology with customizable 25 ports (Mini/Maxi TC options) & bottom Tri-Clamp port for efficient drainage. Plug-and-play integration simplifies setup, reduces contamination risk, and speeds workflows.
Features
Made from USP Class VI materials, gamma-sterilizable, autoclavable (15 PSI, 20 min at 121C)
Rigid dip tube with notch to allow for easy filling and extraction of media
Bottom TC for quick media harvest
Laser marked ports for quick identification
Multi-port cap system
Applications
Bioprocess storage, aseptic transfer
Cell Harvest
Closed collection system
Catalog No. 5000332755 Supplier Foxx Life Sciences Supplier No. 5951001FLS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The EZBio SUT Rigid Reservoir System is a single-use rigid bioprocess carboy and bottle solution for fluid handling, supporting upstream and downstream processes. It provides a sterile, closed, scalable platform with integrated rigid tubing for secure, kink-free transfer of buffers, media, process fluids.
Available in 1L115L volumes (PP bottles/carboys), it features VersaCap technology with customizable 25 ports (Mini/Maxi TC options) & bottom Tri-Clamp port for efficient drainage. Plug-and-play integration simplifies setup, reduces contamination risk, and speeds workflows.
Features
Made from USP Class VI materials, gamma-sterilizable, autoclavable (15 PSI, 20 min at 121C)
Rigid dip tube with notch to allow for easy filling and extraction of media
Bottom TC for quick media harvest
Laser marked ports for quick identification
Multi-port cap system
Applications
Bioprocess storage, aseptic transfer
Cell Harvest
Closed collection system