missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ EZ-Yeast™ Transformation Kit

Catalog No. MP112100200
Change view
Click to view available options
Quantity:
500 μL or 3 to 4 mm colony
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112100200 500 μL or 3 to 4 mm colony
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112100200 Supplier MP Biomedicals™ Supplier No. 112100200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for high-throughput transformation with efficiencies up to 103 transformants per μg plasmid DNA. Perfect for a quick analysis of potentially positive two-hybrid colonies, the EZ-Yeast™ Transformation Kit does not require fully competent cells prior to transformation.

  • The kit does not require fully competent cells prior to transformation
  • Simply remove a 3–4mm colony or 0.5ml overnight culture to a new tube, add vector and carrier DNA, incubate for 30 minutes, and plate on selective media.
  • It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed
     

Specifications

Cell Line S. cerevisiae, S. pombe, S. albicans, many strains
Content And Storage Store EZ-Transformation Solution at Room temperature (15 to 30°C). Store Carrier DNA at 4°C.
Sample Type Yeast Culture
For Use With (Application) High-throughput transformations
No. of Reactions 200 Preps
Quantity 500 μL or 3 to 4 mm colony
Synonym Yeast 2-hybrid reporter kit